CAS RN 135050-44-1
IUPAC Name:
3-chloro-4-iodoaniline
Synonyms:
3-chloro-4-iodoaniline
Molecular Weight (g/mol):
253.47
Molecular Formula:
C6H5ClIN
InChi Key:
ONZHMGRKWJMTDE-UHFFFAOYSA-N
SMILES:
NC1=CC=C(I)C(Cl)=C1
1
–
1
of
1
results
1
3-Chloro-4-iodoaniline 95.0+%, TCI America™
CAS: 135050-44-1 Molecular Formula: C6H5ClIN Molecular Weight (g/mol): 253.47 MDL Number: MFCD00051845 InChI Key: ONZHMGRKWJMTDE-UHFFFAOYSA-N Synonym: benzenamine, 3-chloro-4-iodo,3-chloro-4-iodobenzenamine,4-amino-2-chloroiodobenzene,3-chloro-4-iodophenylamine,pubchem3812,3-chloro4-iodoaniline,zlchem 1106,acmc-209bxp,3-chloro-4-iodo-aniline,3-chloro-4-iodo aniline PubChem CID: 282926 IUPAC Name: 3-chloro-4-iodoaniline SMILES: NC1=CC=C(I)C(Cl)=C1