missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-4-hydroxybenzonitrile 98.0+%, TCI America™
$75.08 - $298.41
Chemical Identifiers
| CAS | 2315-81-3 |
|---|---|
| Molecular Formula | C7H4ClNO |
| Molecular Weight (g/mol) | 153.565 |
| MDL Number | MFCD01567246 |
| InChI Key | CRYPJUOSZDQWJZ-UHFFFAOYSA-N |
| Synonym | benzonitrile, 3-chloro-4-hydroxy, 2-chloro-4-cyanophenol, 3-chloro-4-hydroxy-benzonitrile, 3-chloro-4-hydroxybenzenecarbonitrile, pubchem4726, 4-cyano-2-chlorophenol, buttpark 4357-94, acmc-1clj1, pharmabridge p-1867, buttpark 43\57-94 |
| PubChem CID | 2735739 |
| IUPAC Name | 3-chloro-4-hydroxybenzonitrile |
| SMILES | C1=CC(=C(C=C1C#N)Cl)O |
Chemical Identifiers
| 2315-81-3 | |
| 153.565 | |
| CRYPJUOSZDQWJZ-UHFFFAOYSA-N | |
| 2735739 | |
| C1=CC(=C(C=C1C#N)Cl)O |
| C7H4ClNO | |
| MFCD01567246 | |
| benzonitrile, 3-chloro-4-hydroxy, 2-chloro-4-cyanophenol, 3-chloro-4-hydroxy-benzonitrile, 3-chloro-4-hydroxybenzenecarbonitrile, pubchem4726, 4-cyano-2-chlorophenol, buttpark 4357-94, acmc-1clj1, pharmabridge p-1867, buttpark 43\57-94 | |
| 3-chloro-4-hydroxybenzonitrile |
Specifications
| 2315-81-3 | |
| White-Yellow | |
| MFCD01567246 | |
| benzonitrile, 3-chloro-4-hydroxy, 2-chloro-4-cyanophenol, 3-chloro-4-hydroxy-benzonitrile, 3-chloro-4-hydroxybenzenecarbonitrile, pubchem4726, 4-cyano-2-chlorophenol, buttpark 4357-94, acmc-1clj1, pharmabridge p-1867, buttpark 43\57-94 | |
| C1=CC(=C(C=C1C#N)Cl)O | |
| 153.565 | |
| 153.57 | |
| Crystalline Powder |
| 152°C | |
| C7H4ClNO | |
| 1 g | |
| CRYPJUOSZDQWJZ-UHFFFAOYSA-N | |
| 3-chloro-4-hydroxybenzonitrile | |
| 2735739 | |
| ≥98.0% (GC,T) | |
| 3-Chloro-4-hydroxybenzonitrile |
Safety and Handling
TSCA : No