missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorothiobenzamide, 97%

CAS: 130560-97-3 | C7H5ClFNS | 189.632 g/mol

$83.85 - $204.78

Chemical Identifiers

CAS 130560-97-3
Molecular Formula C7H5ClFNS
Molecular Weight (g/mol) 189.632
MDL Number MFCD00042478
InChI Key RSGPCKCTKDCMRR-UHFFFAOYSA-N
Synonym 3-chloro-4-fluorothiobenzamide, 3-chloro-4-fluorobenzothioamide, 3-chloro-4-fluorobenzene-1-carbothioamide, benzenecarbothioamide,3-chloro-4-fluoro, amino 3-chloro-4-fluorophenyl methane-1-thione, acmc-1ccr0, 3-chloro-4-fluoro-thiobenzamide, #, 3-chloranyl-4-fluoranyl-benzenecarbothioamide, benzenecarbothioamide, 3-chloro-4-fluoro-9ci
PubChem CID 2779521
IUPAC Name 3-chloro-4-fluorobenzenecarbothioamide
SMILES C1=CC(=C(C=C1C(=S)N)Cl)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL11132MD
View Documents
Thermo Scientific Chemicals
L11132MD
250 mg
Each for $83.85
Only null left
 
AAL1113203
View Documents
Thermo Scientific Chemicals
L1113203
1 g
Each for $204.78
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

130560-97-3
189.632
RSGPCKCTKDCMRR-UHFFFAOYSA-N
2779521
C1=CC(=C(C=C1C(=S)N)Cl)F
C7H5ClFNS
MFCD00042478
3-chloro-4-fluorothiobenzamide, 3-chloro-4-fluorobenzothioamide, 3-chloro-4-fluorobenzene-1-carbothioamide, benzenecarbothioamide,3-chloro-4-fluoro, amino 3-chloro-4-fluorophenyl methane-1-thione, acmc-1ccr0, 3-chloro-4-fluoro-thiobenzamide, #, 3-chloranyl-4-fluoranyl-benzenecarbothioamide, benzenecarbothioamide, 3-chloro-4-fluoro-9ci
3-chloro-4-fluorobenzenecarbothioamide

Specifications

130560-97-3
C7H5ClFNS
250 mg
7757675
RSGPCKCTKDCMRR-UHFFFAOYSA-N
3-chloro-4-fluorobenzenecarbothioamide
2779521
97%
129°C to 130°C
MFCD00042478
UN2811
3-chloro-4-fluorothiobenzamide, 3-chloro-4-fluorobenzothioamide, 3-chloro-4-fluorobenzene-1-carbothioamide, benzenecarbothioamide,3-chloro-4-fluoro, amino 3-chloro-4-fluorophenyl methane-1-thione, acmc-1ccr0, 3-chloro-4-fluoro-thiobenzamide, #, 3-chloranyl-4-fluoranyl-benzenecarbothioamide, benzenecarbothioamide, 3-chloro-4-fluoro-9ci
C1=CC(=C(C=C1C(=S)N)Cl)F
189.632
189.64
3-Chloro-4-fluorothiobenzamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only