missing translation for 'onlineSavingsMsg'
Learn More

(3-Chloro-4-fluorophenyl)thiourea 98.0+%, TCI America™

Catalog No. C31371G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31371G 1 g
C31375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31371G Supplier TCI America Supplier No. C31371G

Chemical Identifiers

CAS 154371-25-2
Molecular Formula C7H6ClFN2S
Molecular Weight (g/mol) 204.65
MDL Number MFCD01571081
InChI Key OVJVZVPEMXLKOE-UHFFFAOYSA-N
PubChem CID 704698
IUPAC Name (3-chloro-4-fluorophenyl)thiourea
SMILES NC(=S)NC1=CC(Cl)=C(F)C=C1

Specifications

Melting Point 180°C
Color White-Yellow
Quantity 1 g
Formula Weight 204.65
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (3-Chloro-4-fluorophenyl)thiourea