accessibility menu, dialog, popup

UserName

CAS RN 154371-25-2

IUPAC Name: (3-chloro-4-fluorophenyl)thiourea
Synonyms: 3-chloro-4-fluorophenylthiourea
Molecular Weight (g/mol): 204.65
Molecular Formula: C7H6ClFN2S
InChi Key: OVJVZVPEMXLKOE-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC(Cl)=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results