missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorophenyl isocyanate, 98%

Catalog No. AAL1089106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1089106 5 g
AAL1089114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1089106 Supplier Thermo Scientific Chemicals Supplier No. L1089106
Only null left

CAS: 50529-33-4 | C7H3ClFNO | 171.555 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50529-33-4
Molecular Formula C7H3ClFNO
Molecular Weight (g/mol) 171.555
MDL Number MFCD00037037
InChI Key XVIPJBUXMFLHSI-UHFFFAOYSA-N
Synonym 3-chloro-4-fluorophenyl isocyanate, 3-chloro-4-fluorophenylisocyanate, benzene, 2-chloro-1-fluoro-4-isocyanato, 3-chloro-4-fluorobenzenisocyanate, pubchem5024, acmc-20an6r, 3-chloro-4-fluoro-phenylisocyanate, 3-chloro-4-fluorophenyl-isocyanate, 3-chloro-4-fluoro phenyl isocyanate
PubChem CID 2734632
IUPAC Name 2-chloro-1-fluoro-4-isocyanatobenzene
SMILES C1=CC(=C(C=C1N=C=O)Cl)F

Specifications

Density 1.377
Boiling Point 44°C (17 mmHg)
Flash Point 87°C (188°F)
Refractive Index 1.541
Quantity 5 g
UN Number UN2206
Beilstein 973324
Sensitivity Moisture sensitive
Formula Weight 171.56
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-fluorophenyl isocyanate
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H300-H330-H334-H227-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only