CAS RN 50529-33-4
IUPAC Name:
2-chloro-1-fluoro-4-isocyanatobenzene
Synonyms:
2-chloro-1-fluoro-4-isocyanatobenzene
Molecular Weight (g/mol):
171.56
Molecular Formula:
C7H3ClFNO
InChi Key:
XVIPJBUXMFLHSI-UHFFFAOYSA-N
SMILES:
FC1=CC=C(C=C1Cl)N=C=O
1
–
1
of
1
results
1
Promotions Available
3-Chloro-4-fluorophenyl isocyanate, 98%
3-Chloro-4-fluorophenyl isocyanate, 98%
CAS: 50529-33-4 Molecular Formula: C7H3ClFNO Molecular Weight (g/mol): 171.555 MDL Number: MFCD00037037 InChI Key: XVIPJBUXMFLHSI-UHFFFAOYSA-N Synonym: 3-chloro-4-fluorophenyl isocyanate,3-chloro-4-fluorophenylisocyanate,benzene, 2-chloro-1-fluoro-4-isocyanato,3-chloro-4-fluorobenzenisocyanate,pubchem5024,acmc-20an6r,3-chloro-4-fluoro-phenylisocyanate,3-chloro-4-fluorophenyl-isocyanate,3-chloro-4-fluoro phenyl isocyanate PubChem CID: 2734632 IUPAC Name: 2-chloro-1-fluoro-4-isocyanatobenzene SMILES: C1=CC(=C(C=C1N=C=O)Cl)F