missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-4-fluorobenzoyl Chloride 98.0+%, TCI America™
Supplier: TCI America C212225G
Specifications
| 3-Chloro-4-fluorobenzoyl Chloride | |
| Yellow | |
| C7H3Cl2FO | |
| 25 g | |
| 4-fluoro-3-chlorobenzoyl chloride, 3-chloro-4-fluorobenzoylchloride, 3-chloro-4-fluoro benzoyl chloride, 3-chloro-4-fluoro-benzoyl chloride, benzoyl chloride, 3-chloro-4-fluoro, acmc-209np6, dsstox_cid_29301, dsstox_rid_83419, dsstox_gsid_49344, ksc622a8f | |
| C1=CC(=C(C=C1C(=O)Cl)Cl)F | |
| 192.998 | |
| 193.00 | |
| Liquid |
| 65055-17-6 | |
| 222°C | |
| MFCD00236215 | |
| 3265 | |
| DLZUHSFUBZTBQZ-UHFFFAOYSA-N | |
| 3-chloro-4-fluorobenzoyl chloride | |
| 2736548 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 65055-17-6 | |
| 192.998 | |
| DLZUHSFUBZTBQZ-UHFFFAOYSA-N | |
| 2736548 | |
| C1=CC(=C(C=C1C(=O)Cl)Cl)F |
| C7H3Cl2FO | |
| MFCD00236215 | |
| 4-fluoro-3-chlorobenzoyl chloride, 3-chloro-4-fluorobenzoylchloride, 3-chloro-4-fluoro benzoyl chloride, 3-chloro-4-fluoro-benzoyl chloride, benzoyl chloride, 3-chloro-4-fluoro, acmc-209np6, dsstox_cid_29301, dsstox_rid_83419, dsstox_gsid_49344, ksc622a8f | |
| 3-chloro-4-fluorobenzoyl chloride |
Safety and Handling
TSCA : No