missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorobenzoyl Chloride 98.0+%, TCI America™

Catalog No. p-7125294
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21225G 5 g
C212225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21225G Supplier TCI America Supplier No. C21225G

Chemical Identifiers

CAS 65055-17-6
Molecular Formula C7H3Cl2FO
Molecular Weight (g/mol) 192.998
MDL Number MFCD00236215
InChI Key DLZUHSFUBZTBQZ-UHFFFAOYSA-N
Synonym 4-fluoro-3-chlorobenzoyl chloride, 3-chloro-4-fluorobenzoylchloride, 3-chloro-4-fluoro benzoyl chloride, 3-chloro-4-fluoro-benzoyl chloride, benzoyl chloride, 3-chloro-4-fluoro, acmc-209np6, dsstox_cid_29301, dsstox_rid_83419, dsstox_gsid_49344, ksc622a8f
PubChem CID 2736548
IUPAC Name 3-chloro-4-fluorobenzoyl chloride
SMILES C1=CC(=C(C=C1C(=O)Cl)Cl)F

Specifications

Color Yellow
Boiling Point 222°C
Quantity 5 g
UN Number 3265
Formula Weight 193.00
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Chloro-4-fluorobenzoyl Chloride