missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorobenzotrifluoride 98.0+%, TCI America™

Catalog No. p-7125030
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
C15155G 5 g
C151525G 25 g
2 options

Catalog No. C15155G

Supplier: TCI America C15155G

Only null left

Chemical Identifiers

CAS 78068-85-6
Molecular Formula C7H3ClF4
Molecular Weight (g/mol) 198.54
MDL Number MFCD00042207
InChI Key BKHVEYHSOXVAOP-UHFFFAOYSA-N
Synonym 3-chloro-4-fluorobenzotrifluoride, 2-chloro-1-fluoro-4-trifluoromethyl benzene, benzene, 2-chloro-1-fluoro-4-trifluoromethyl, 3-cholor-4-fluorobenzo-trifluoride, 3-chloro-4-fluoro benzotrifluoride, 1-chloro-2-fluoro-5-trifluoromethyl benzene, 3-chloro-4-fluoro-1-trifluoromethyl benzene, benzene, chloro-1-fluoro-4-trifluoromethyl, 3-chloro-alpha,alpha,alpha,4-tetrafluorotoluene, 3-chloro-4-fluoro-alpha,alpha,alpha-trifluorotoluene
PubChem CID 157153
IUPAC Name 2-chloro-1-fluoro-4-(trifluoromethyl)benzene
SMILES FC1=CC=C(C=C1Cl)C(F)(F)F

Specifications

Quantity 5 g
Formula Weight 198.54
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-4-fluorobenzotrifluoride
EINECSNumber (3)-4051
TSCA Yes
Recommended Storage Refrigerator