accessibility menu, dialog, popup

UserName

CAS RN 78068-85-6

IUPAC Name: 2-chloro-1-fluoro-4-(trifluoromethyl)benzene
Synonyms: 2-chloro-1-fluoro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 198.54
Molecular Formula: C7H3ClF4
InChi Key: BKHVEYHSOXVAOP-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1Cl)C(F)(F)F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results