missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorobenzotrifluoride, 98%

CAS: 78068-85-6 | C7H3ClF4 | 198.54 g/mol

Supplier:  Thermo Scientific Chemicals A1667006

 View more versions of this product

Catalog No. AAA1667006


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Chloro-4-fluorobenzotrifluoride
1.49
42°C (108°F)
1.4355
5 g
4309386
BKHVEYHSOXVAOP-UHFFFAOYSA-N
2-chloro-1-fluoro-4-(trifluoromethyl)benzene
157153
98%
78068-85-6
137°C
C7H3ClF4
MFCD00042207
UN1993
3-chloro-4-fluorobenzotrifluoride, 2-chloro-1-fluoro-4-trifluoromethyl benzene, benzene, 2-chloro-1-fluoro-4-trifluoromethyl, 3-cholor-4-fluorobenzo-trifluoride, 3-chloro-4-fluoro benzotrifluoride, 1-chloro-2-fluoro-5-trifluoromethyl benzene, 3-chloro-4-fluoro-1-trifluoromethyl benzene, benzene, chloro-1-fluoro-4-trifluoromethyl, 3-chloro-alpha,alpha,alpha,4-tetrafluorotoluene, 3-chloro-4-fluoro-alpha,alpha,alpha-trifluorotoluene
FC1=CC=C(C=C1Cl)C(F)(F)F
198.54
198.55

Chemical Identifiers

78068-85-6
198.54
BKHVEYHSOXVAOP-UHFFFAOYSA-N
157153
FC1=CC=C(C=C1Cl)C(F)(F)F
C7H3ClF4
MFCD00042207
3-chloro-4-fluorobenzotrifluoride, 2-chloro-1-fluoro-4-trifluoromethyl benzene, benzene, 2-chloro-1-fluoro-4-trifluoromethyl, 3-cholor-4-fluorobenzo-trifluoride, 3-chloro-4-fluoro benzotrifluoride, 1-chloro-2-fluoro-5-trifluoromethyl benzene, 3-chloro-4-fluoro-1-trifluoromethyl benzene, benzene, chloro-1-fluoro-4-trifluoromethyl, 3-chloro-alpha,alpha,alpha,4-tetrafluorotoluene, 3-chloro-4-fluoro-alpha,alpha,alpha-trifluorotoluene
2-chloro-1-fluoro-4-(trifluoromethyl)benzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only