missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorobenzaldehyde 98.0+%, TCI America™

Catalog No. p-7125017
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C14985G 5 g
C149825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C14985G Supplier TCI America Supplier No. C14985G

Chemical Identifiers

CAS 34328-61-5
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.56
MDL Number MFCD00011735
InChI Key GVORVQPNNSASDM-UHFFFAOYSA-N
Synonym 3-chloro-4-fluoro-benzaldehyde, benzaldehyde, 3-chloro-4-fluoro, 3-chloro-4-fluorobenzaldehyde,, pubchem1409, acmc-1bmpv, 3-chloro4-fluorobenzaldehyde, ksc496c9d, 3-chloro-4fluoro-benzaldehyde, 4-fluoro-3-chlorobenzaldehyde, 3-chloro,4-fluorobenzaldehyde;
PubChem CID 593866
IUPAC Name 3-chloro-4-fluorobenzaldehyde
SMILES FC1=CC=C(C=O)C=C1Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 158.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Chloro-4-fluorobenzaldehyde