accessibility menu, dialog, popup

UserName

FOCl

CAS RN 34328-61-5

IUPAC Name: 3-chloro-4-fluorobenzaldehyde
Synonyms: 3-chloro-4-fluorobenzaldehyde
Molecular Weight (g/mol): 158.56
Molecular Formula: C7H4ClFO
InChi Key: GVORVQPNNSASDM-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=O)C=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results