missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluoro-5-nitrobenzotrifluoride, 98%, Thermo Scientific Chemicals

Catalog No. AAL1932206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1932206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1932206 Supplier Thermo Scientific Chemicals Supplier No. L1932206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 101646-02-0 | Molecular Formula: C7H2ClF4NO2 | Molecular Weight: 243.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 101646-02-0
Molecular Formula C7H2ClF4NO2
Molecular Weight (g/mol) 243.54
MDL Number MFCD00674108
InChI Key JVOKKJHIEVEFEB-UHFFFAOYSA-N
Synonym 3-chloro-4-fluoro-5-nitrobenzotrifluoride, 1-chloro-2-fluoro-3-nitro-5-trifluoromethyl benzene, 3-chloro-2-fluoro-1-nitro-5-trifluoromethyl benzene, 5-chloro-4-fluoro-3-nitrotrifluoromethylbenzene, 5-chloro-4-fluoro-3-nitro-trifluoromethylbenzene
PubChem CID 2736565
IUPAC Name 1-chloro-2-fluoro-3-nitro-5-(trifluoromethyl)benzene
SMILES [O-][N+](=O)C1=C(F)C(Cl)=CC(=C1)C(F)(F)F

Specifications

Density 1.61
Boiling Point 200°C
Flash Point >110°C (230°F)
Refractive Index 1.482
Quantity 5 g
UN Number UN2810
Beilstein 5436538
Formula Weight 243.55
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-fluoro-5-nitrobenzotrifluoride
Hazard Category H302+H332-H311-H315-H319
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only