missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-methylaniline Hydrochloride 98.0+%, TCI America™

Catalog No. A028525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A028525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A028525G Supplier TCI America Supplier No. A028525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6259-40-1
Molecular Formula C7H9Cl2N
Molecular Weight (g/mol) 178.056
MDL Number MFCD00058279
InChI Key WCZGXAZCBNLQJB-UHFFFAOYSA-N
Synonym 2-Amino-6-chlorotoluene Hydrochloride, 3-Chloro-o-toluidine Hydrochloride
PubChem CID 80415
IUPAC Name 3-chloro-2-methylaniline;hydrochloride
SMILES CC1=C(C=CC=C1Cl)N.Cl

Specifications

Quantity 25 g
Formula Weight 178.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-2-methylaniline Hydrochloride
EINECSNumber (3)-0265
TSCA Yes