accessibility menu, dialog, popup

UserName

CAS RN 6259-40-1

IUPAC Name: hydrogen 3-chloro-2-methylaniline chloride
Synonyms: hydrogen 2-methyl-3-chloroaniline chloride
Molecular Weight (g/mol): 178.06
Molecular Formula: C7H9Cl2N
InChi Key: WCZGXAZCBNLQJB-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=C(N)C=CC=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results