missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-2,4-difluorobenzoic Acid 98.0+%, TCI America™
Supplier: TCI America C251725G
Specifications
| 3-Chloro-2,4-difluorobenzoic Acid | |
| 176°C | |
| MFCD01631387 | |
| 3-chloro-2,4-difluoro-benzoic acid, 2,4-difluoro-3-chlorobenzoic acid, 3-chloro-2,4-difluorobenzoicacid, benzoic acid, 3-chloro-2,4-difluoro, pubchem2217, acmc-1byme, intermediates-zcf02173, ksc181k3p | |
| [O-]C(=O)C1=CC=C(F)C(Cl)=C1F | |
| 191.54 | |
| 192.55 | |
| Crystalline Powder |
| 154257-75-7 | |
| C7H2ClF2O2 | |
| 25 g | |
| YGYZTZAEZMCPSC-UHFFFAOYSA-M | |
| 3-chloro-2,4-difluorobenzoate | |
| 2773532 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 154257-75-7 | |
| 191.54 | |
| YGYZTZAEZMCPSC-UHFFFAOYSA-M | |
| 2773532 | |
| [O-]C(=O)C1=CC=C(F)C(Cl)=C1F |
| C7H2ClF2O2 | |
| MFCD01631387 | |
| 3-chloro-2,4-difluoro-benzoic acid, 2,4-difluoro-3-chlorobenzoic acid, 3-chloro-2,4-difluorobenzoicacid, benzoic acid, 3-chloro-2,4-difluoro, pubchem2217, acmc-1byme, intermediates-zcf02173, ksc181k3p | |
| 3-chloro-2,4-difluorobenzoate |
Safety and Handling
TSCA : No