missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2,4-difluorobenzoic Acid 98.0+%, TCI America™

Catalog No. p-7125504
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25175G 5 g
C251725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25175G Supplier TCI America Supplier No. C25175G

Chemical Identifiers

CAS 154257-75-7
Molecular Formula C7H2ClF2O2
Molecular Weight (g/mol) 191.54
MDL Number MFCD01631387
InChI Key YGYZTZAEZMCPSC-UHFFFAOYSA-M
Synonym 3-chloro-2,4-difluoro-benzoic acid, 2,4-difluoro-3-chlorobenzoic acid, 3-chloro-2,4-difluorobenzoicacid, benzoic acid, 3-chloro-2,4-difluoro, pubchem2217, acmc-1byme, intermediates-zcf02173, ksc181k3p
PubChem CID 2773532
IUPAC Name 3-chloro-2,4-difluorobenzoate
SMILES [O-]C(=O)C1=CC=C(F)C(Cl)=C1F

Specifications

Melting Point 176°C
Quantity 5 g
Formula Weight 192.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-2,4-difluorobenzoic Acid