missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1,2-benzisothiazole, 97+%

CAS: 7716-66-7 | C7H4ClNS | 169.626 g/mol

Supplier:  Thermo Scientific Chemicals L1779114

 View more versions of this product

Catalog No. AAL1779114

Only null left
Explore available promotions

Description

Description

3-Chloro-1,2-benzisothiazole is used as an intermediate of Ziprasidone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-1,2-benzisothiazole is used as an intermediate of Ziprasidone.

Solubility
Soluble in Chloroform, Dichloromethane and Ethyl Acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications

Specifications

3-Chloro-1,2-benzisothiazole
37°C to 44°C
C7H4ClNS
25 g
3-chloro-1,2-benzisothiazole, 3-chlorobenzo d isothiazole, 3-chlorobenzisothiazole, 3-chloro-1,2-benzoisothiazole, 1,2-benzisothiazole, 3-chloro, 3-chloro-benzo d isothiazole, chlorobenzisothiazole, pubchem22324, acmc-1bdko, 3-chlorobenzo d isothia-zole
BCPVKLRBQLRWDQ-UHFFFAOYSA-N
3-chloro-1,2-benzothiazole
598190
99%
7716-66-7
80°C to 86°C (0.75 mmHg)
MFCD00673254
119370
Soluble in Chloroform,Dichloromethane and Ethyl Acetate.
C1=CC=C2C(=C1)C(=NS2)Cl
169.626
169.63

Chemical Identifiers

7716-66-7
169.626
BCPVKLRBQLRWDQ-UHFFFAOYSA-N
598190
C1=CC=C2C(=C1)C(=NS2)Cl
C7H4ClNS
MFCD00673254
3-chloro-1,2-benzisothiazole, 3-chlorobenzo d isothiazole, 3-chlorobenzisothiazole, 3-chloro-1,2-benzoisothiazole, 1,2-benzisothiazole, 3-chloro, 3-chloro-benzo d isothiazole, chlorobenzisothiazole, pubchem22324, acmc-1bdko, 3-chlorobenzo d isothia-zole
3-chloro-1,2-benzothiazole
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only