missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-1,2-benzisothiazole, 97+%

Catalog No. p-7047153
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1779106 5 g
AAL1779114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1779106 Supplier Thermo Scientific Chemicals Supplier No. L1779106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7716-66-7 | C7H4ClNS | 169.626 g/mol

3-Chloro-1,2-benzisothiazole is used as an intermediate of Ziprasidone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-1,2-benzisothiazole is used as an intermediate of Ziprasidone.

Solubility
Soluble in Chloroform, Dichloromethane and Ethyl Acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 7716-66-7
Molecular Formula C7H4ClNS
Molecular Weight (g/mol) 169.626
MDL Number MFCD00673254
InChI Key BCPVKLRBQLRWDQ-UHFFFAOYSA-N
Synonym 3-chloro-1,2-benzisothiazole, 3-chlorobenzo d isothiazole, 3-chlorobenzisothiazole, 3-chloro-1,2-benzoisothiazole, 1,2-benzisothiazole, 3-chloro, 3-chloro-benzo d isothiazole, chlorobenzisothiazole, pubchem22324, acmc-1bdko, 3-chlorobenzo d isothia-zole
PubChem CID 598190
IUPAC Name 3-chloro-1,2-benzothiazole
SMILES C1=CC=C2C(=C1)C(=NS2)Cl

Specifications

Melting Point 37°C to 44°C
Boiling Point 80°C to 86°C (0.75 mmHg)
Quantity 5 g
Beilstein 119370
Solubility Information Soluble in Chloroform,Dichloromethane and Ethyl Acetate.
Formula Weight 169.63
Percent Purity 99%
Chemical Name or Material 3-Chloro-1,2-benzisothiazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only