accessibility menu, dialog, popup

UserName

CAS RN 7716-66-7

IUPAC Name: 3-chloro-1,2-benzothiazole
Synonyms: 3-chloro-1,2-benzothiazole
Molecular Weight (g/mol): 169.63
Molecular Formula: C7H4ClNS
InChi Key: BCPVKLRBQLRWDQ-UHFFFAOYSA-N
SMILES: ClC1=NSC2=CC=CC=C12
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)