Learn More
(3-Carboxypropyl)triphenylphosphonium bromide, 97%
CAS: 17857-14-6 | C22H22BrO2P | 429.29 g/mol
$124.81 - $1274.55
Chemical Identifiers
| CAS | 17857-14-6 |
|---|---|
| Molecular Formula | C22H22BrO2P |
| Molecular Weight (g/mol) | 429.29 |
| MDL Number | MFCD00274196 |
| InChI Key | NKVJKVMGJABKHV-UHFFFAOYSA-N |
| Synonym | 3-carboxypropyltriphenylphosphonium bromide, 3-carboxypropyl triphenylphosphonium bromide, 3-carboxypropyl triphenylphosphanium bromide, carboxypropyltriphenylphosphoniumbromide, phosphonium, 3-carboxypropyl triphenyl-, bromide, 4-hydroxy-4-oxobutyl triphenylphosphonium bromide, 3-carboxypropyl triphenylphosphonium bromide 3-carboxypropyl triphenylphosphonium bromide, acmc-1bp1u, xfzdcviyhhhybq-uhfffaoysa-m |
| PubChem CID | 10717451 |
| IUPAC Name | 3-carboxypropyl(triphenyl)phosphanium;bromide |
| SMILES | [Br-].OC(=O)CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1930214
|
Thermo Scientific Chemicals
A1930214 |
25 g |
Each for $124.81
|
|
|||||
|
AAA1930222
|
Thermo Scientific Chemicals
A1930222 |
100 g |
Each for $340.70
|
|
|||||
|
AAA1930236
|
Thermo Scientific Chemicals
A1930236 |
500 g |
Each for $1,274.55
|
|
|||||
Description
Reactant for preparation of Piperamide analogs as histone deacetylase (HDAC) inhibitors with antitumor activity, Boron-containing benzoxaboroles as antimalarial agents, Methyl alkenyl quinolones as antimycobacterial agents, Solandelactone E via Sharpless epoxidation, Taber cyclopropanation, chemoselective reductions and lithiation-borylation-allylation sequence, Organotin compounds as antifungal agents, Oleanolic acid derivatives as inhibitors of protein tyrosine phosphatase 1B for negative regulation of insulin pathway (a promising target for treatment of diabetes and obesity), 11-oxa prostaglandin analogs with ocular hypotensive activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReactant for preparation of Piperamide analogs as histone deacetylase (HDAC) inhibitors with antitumor activity, Boron-containing benzoxaboroles as antimalarial agents, Methyl alkenyl quinolones as antimycobacterial agents, Solandelactone E via Sharpless epoxidation, Taber cyclopropanation, chemoselective reductions and lithiation-borylation-allylation sequence, Organotin compounds as antifungal agents, Oleanolic acid derivatives as inhibitors of protein tyrosine phosphatase 1B for negative regulation of insulin pathway (a promising target for treatment of diabetes and obesity), 11-oxa prostaglandin analogs with ocular hypotensive activity.
Solubility
Soluble in water. soluble in ethanol.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Chemical Identifiers
| 17857-14-6 | |
| 429.29 | |
| NKVJKVMGJABKHV-UHFFFAOYSA-N | |
| 10717451 | |
| [Br-].OC(=O)CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
| C22H22BrO2P | |
| MFCD00274196 | |
| 3-carboxypropyltriphenylphosphonium bromide, 3-carboxypropyl triphenylphosphonium bromide, 3-carboxypropyl triphenylphosphanium bromide, carboxypropyltriphenylphosphoniumbromide, phosphonium, 3-carboxypropyl triphenyl-, bromide, 4-hydroxy-4-oxobutyl triphenylphosphonium bromide, 3-carboxypropyl triphenylphosphonium bromide 3-carboxypropyl triphenylphosphonium bromide, acmc-1bp1u, xfzdcviyhhhybq-uhfffaoysa-m | |
| 3-carboxypropyl(triphenyl)phosphanium;bromide |
Specifications
| 17857-14-6 | |
| C22H22BrO2P | |
| 25 g | |
| Hygroscopic | |
| Soluble in water. soluble in ethanol. | |
| [Br-].OC(=O)CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 429.29 | |
| 429.3 | |
| (3-Carboxypropyl)triphenylphosphonium bromide |
| 244°C to 250°C | |
| MFCD00274196 | |
| 4173599 | |
| 3-carboxypropyltriphenylphosphonium bromide, 3-carboxypropyl triphenylphosphonium bromide, 3-carboxypropyl triphenylphosphanium bromide, carboxypropyltriphenylphosphoniumbromide, phosphonium, 3-carboxypropyl triphenyl-, bromide, 4-hydroxy-4-oxobutyl triphenylphosphonium bromide, 3-carboxypropyl triphenylphosphonium bromide 3-carboxypropyl triphenylphosphonium bromide, acmc-1bp1u, xfzdcviyhhhybq-uhfffaoysa-m | |
| NKVJKVMGJABKHV-UHFFFAOYSA-N | |
| 3-carboxypropyl(triphenyl)phosphanium;bromide | |
| 10717451 | |
| 97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only