missing translation for 'onlineSavingsMsg'
Learn More

3-Carboxy-5-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog No. AAH6222503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6222503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6222503 Supplier Thermo Scientific Chemicals Supplier No. H6222503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 936728-22-2
Molecular Formula C13H16BFO4
Molecular Weight (g/mol) 266.075
MDL Number MFCD12756465
InChI Key NMYIHYHALOHVHV-UHFFFAOYSA-N
Synonym 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxy-5-fluorophenylboronic acid, pinacol ester, 3-carboxy-5-fluorobenzeneboronic acid pinacol ester, 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoicacid, 3-fluoro-5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid
PubChem CID 46739807
IUPAC Name 3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)F)C(=O)O

Specifications

Quantity 1 g
Formula Weight 266.07
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Carboxy-5-fluorobenzeneboronic acid pinacol ester
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only