CAS RN 936728-22-2
IUPAC Name:
3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Synonyms:
3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Molecular Weight (g/mol):
266.08
Molecular Formula:
C13H16BFO4
InChi Key:
NMYIHYHALOHVHV-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=CC(=CC(F)=C1)C(O)=O
1
–
1
of
1
results
1
Promotions Available
3-Carboxy-5-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
3-Carboxy-5-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals
CAS: 936728-22-2 Molecular Formula: C13H16BFO4 Molecular Weight (g/mol): 266.075 MDL Number: MFCD12756465 InChI Key: NMYIHYHALOHVHV-UHFFFAOYSA-N Synonym: 3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid,3-carboxy-5-fluorophenylboronic acid, pinacol ester,3-carboxy-5-fluorobenzeneboronic acid pinacol ester,3-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoicacid,3-fluoro-5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid PubChem CID: 46739807 IUPAC Name: 3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid SMILES: B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)F)C(=O)O