missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Buten-2-yl Acetate 98.0+%, TCI America™
$492.11 - $1959.13
Chemical Identifiers
| CAS | 6737-11-7 |
|---|---|
| Molecular Formula | C6H10O2 |
| Molecular Weight (g/mol) | 114.14 |
| MDL Number | MFCD00026202 |
| InChI Key | LYWCPTCPTWCZSZ-UHFFFAOYNA-N |
| Synonym | 3-Acetoxy-1-butene, Acetic Acid 3-Buten-2-yl Ester, 1-Methylallyl Acetate |
| PubChem CID | 138821 |
| IUPAC Name | but-3-en-2-yl acetate |
| SMILES | CC(C=C)OC(=O)C |
Chemical Identifiers
| 6737-11-7 | |
| 114.14 | |
| LYWCPTCPTWCZSZ-UHFFFAOYNA-N | |
| 138821 | |
| CC(C=C)OC(=O)C |
| C6H10O2 | |
| MFCD00026202 | |
| 3-Acetoxy-1-butene, Acetic Acid 3-Buten-2-yl Ester, 1-Methylallyl Acetate | |
| but-3-en-2-yl acetate |
Specifications
| 6737-11-7 | |
| 112°C | |
| MFCD00026202 | |
| 3272 | |
| LYWCPTCPTWCZSZ-UHFFFAOYNA-N | |
| but-3-en-2-yl acetate | |
| 138821 | |
| ≥98.0% (GC) | |
| 3-Buten-2-yl Acetate |
| Yellow | |
| C6H10O2 | |
| 5 g | |
| 3-Acetoxy-1-butene, Acetic Acid 3-Buten-2-yl Ester, 1-Methylallyl Acetate | |
| CC(C=C)OC(=O)C | |
| 114.14 | |
| 114.14 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0760
TSCA : No