missing translation for 'onlineSavingsMsg'
Learn More

3-Buten-2-yl Acetate 98.0+%, TCI America™

Supplier:  TCI America B01285G

 View more versions of this product

Catalog No. B01285G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Buten-2-yl Acetate
Yellow
C6H10O2
5 g
3-Acetoxy-1-butene, Acetic Acid 3-Buten-2-yl Ester, 1-Methylallyl Acetate
CC(C=C)OC(=O)C
114.14
114.14
Liquid
6737-11-7
112°C
MFCD00026202
3272
LYWCPTCPTWCZSZ-UHFFFAOYNA-N
but-3-en-2-yl acetate
138821
≥98.0% (GC)

Chemical Identifiers

6737-11-7
114.14
LYWCPTCPTWCZSZ-UHFFFAOYNA-N
138821
CC(C=C)OC(=O)C
C6H10O2
MFCD00026202
3-Acetoxy-1-butene, Acetic Acid 3-Buten-2-yl Ester, 1-Methylallyl Acetate
but-3-en-2-yl acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0760

TSCA : No

SDS
Documents

Documents

Promotions

Promotions