missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Buten-2-yl Acetate 98.0+%, TCI America™
Supplier: TCI America B012825G
Specifications
| 3-Buten-2-yl Acetate | |
| Yellow | |
| C6H10O2 | |
| 25 g | |
| 3-Acetoxy-1-butene, Acetic Acid 3-Buten-2-yl Ester, 1-Methylallyl Acetate | |
| CC(C=C)OC(=O)C | |
| 114.14 | |
| 114.14 | |
| Liquid |
| 6737-11-7 | |
| 112°C | |
| MFCD00026202 | |
| 3272 | |
| LYWCPTCPTWCZSZ-UHFFFAOYNA-N | |
| but-3-en-2-yl acetate | |
| 138821 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 6737-11-7 | |
| 114.14 | |
| LYWCPTCPTWCZSZ-UHFFFAOYNA-N | |
| 138821 | |
| CC(C=C)OC(=O)C |
| C6H10O2 | |
| MFCD00026202 | |
| 3-Acetoxy-1-butene, Acetic Acid 3-Buten-2-yl Ester, 1-Methylallyl Acetate | |
| but-3-en-2-yl acetate |
Safety and Handling
EINECSNumber : (2)-0760
TSCA : No