missing translation for 'onlineSavingsMsg'
Learn More

3-Bromotoluene 98.0+%, TCI America™

Supplier:  TCI America B0660500G

 View more versions of this product

Catalog No. B0660500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Bromotoluene
Yellow
C7H7Br
500 g
3-bromotoluene, m-bromotoluene, 3-methylbromobenzene, benzene, 1-bromo-3-methyl, m-tolyl bromide, toluene, m-bromo, 5-bromotoluene, 3-bromo-1-methylbenzene, m-methylbromobenzene, 3-methyl-1-bromobenzene
CC1=CC=CC(Br)=C1
171.04
171.04
Liquid
591-17-3
184°C
MFCD00000085
1993
WJIFKOVZNJTSGO-UHFFFAOYSA-N
1-bromo-3-methylbenzene
11560
≥98.0% (GC)

Chemical Identifiers

591-17-3
171.04
WJIFKOVZNJTSGO-UHFFFAOYSA-N
11560
CC1=CC=CC(Br)=C1
C7H7Br
MFCD00000085
3-bromotoluene, m-bromotoluene, 3-methylbromobenzene, benzene, 1-bromo-3-methyl, m-tolyl bromide, toluene, m-bromo, 5-bromotoluene, 3-bromo-1-methylbenzene, m-methylbromobenzene, 3-methyl-1-bromobenzene
1-bromo-3-methylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0088

RTECSNumber : XS7965400

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions