accessibility menu, dialog, popup

UserName

CAS RN 591-17-3

IUPAC Name: 1-bromo-3-methylbenzene
Synonyms: M-bromotoluene
Molecular Weight (g/mol): 171.04
Molecular Formula: C7H7Br
InChi Key: WJIFKOVZNJTSGO-UHFFFAOYSA-N
SMILES: CC1=CC=CC(Br)=C1
Boiling Point 
  • (3)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (5)