missing translation for 'onlineSavingsMsg'
Learn More

3-Bromotoluene 98.0+%, TCI America™

Catalog No. B066025G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B066025G 25 g
B0660100G 100 g
B0660500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B066025G Supplier TCI America Supplier No. B066025G

Chemical Identifiers

CAS 591-17-3
Molecular Formula C7H7Br
Molecular Weight (g/mol) 171.04
MDL Number MFCD00000085
InChI Key WJIFKOVZNJTSGO-UHFFFAOYSA-N
Synonym 3-bromotoluene, m-bromotoluene, 3-methylbromobenzene, benzene, 1-bromo-3-methyl, m-tolyl bromide, toluene, m-bromo, 5-bromotoluene, 3-bromo-1-methylbenzene, m-methylbromobenzene, 3-methyl-1-bromobenzene
PubChem CID 11560
IUPAC Name 1-bromo-3-methylbenzene
SMILES CC1=CC=CC(Br)=C1

Specifications

Color Yellow
Boiling Point 184°C
Quantity 25 g
UN Number 1993
Formula Weight 171.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromotoluene
EINECSNumber (3)-0088
RTECSNumber XS7965400
TSCA Yes