missing translation for 'onlineSavingsMsg'
Learn More

3-Bromotoluene, 98%, Thermo Scientific Chemicals

Catalog No. p-77308
Change view
Click to view available options
Quantity:
10 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC173410100 10 mL Glass bottle
AC173411000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC173410100 Supplier Thermo Scientific Chemicals Supplier No. 173410100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 591-17-3 | C7H7Br | 171.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 591-17-3
Molecular Formula C7H7Br
Molecular Weight (g/mol) 171.04
MDL Number MFCD00000085
InChI Key WJIFKOVZNJTSGO-UHFFFAOYSA-N
Synonym 3-bromotoluene, m-bromotoluene, 3-methylbromobenzene, benzene, 1-bromo-3-methyl, m-tolyl bromide, toluene, m-bromo, 5-bromotoluene, 3-bromo-1-methylbenzene, m-methylbromobenzene, 3-methyl-1-bromobenzene
PubChem CID 11560
IUPAC Name 1-bromo-3-methylbenzene
SMILES CC1=CC=CC(Br)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -40.0°C
Color Colorless to Yellow
Density 1.4100g/mL
Boiling Point 183.0°C to 184.0°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5510 to 1.5530
Linear Formula CH3C6H4Br
Quantity 10 mL
Beilstein 05, 305
Specific Gravity 1.41
Merck Index 14, 1439
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether and benzene
Formula Weight 171.04
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Bromotoluene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
EINECSNumber 209-702-3
RTECSNumber XS7965400
TSCA TSCA

RUO – Research Use Only