Learn More
3-Bromotoluene, 98%
Description
3-Bromotoluene was used in the precise determination of bromine isotope ratio in organic compounds by MC-ICPMS ( Multicollector-Inductively Coupled Plasma Mass Spectrometer). It undergoes palladium-catalyzed cyanation reaction in the presence of K4[Fe(CN)6] as the cyanide surrogate. It also undergoes palladium-catalyzed reaction with alkynyltriarylborates to yield trisubstituted alkenylboranes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Bromotoluene was used in the precise determination of bromine isotope ratio in organic compounds by MC-ICPMS ( Multicollector-Inductively Coupled Plasma Mass Spectrometer). It undergoes palladium-catalyzed cyanation reaction in the presence of K4[Fe(CN)6] as the cyanide surrogate. It also undergoes palladium-catalyzed reaction with alkynyltriarylborates to yield trisubstituted alkenylboranes.
Solubility
Not miscible in water. Soluble in alcohols, ether and benzene.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | -40°C |
| Density | 1.41 |
| Boiling Point | 183°C to 184°C |
| Flash Point | 75°C (167°F) |
| Refractive Index | 1.552 |
| Quantity | 500 g |
| Beilstein | 1903633 |
| Merck Index | 14,1439 |
| Solubility Information | Not miscible in water. Soluble in alcohols,ether and benzene. |
| Formula Weight | 171.04 |
| Show More |
Safety and Handling
| Hazard Category | H226-H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335-H227 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 3; Packaging Group: III |
| EINECSNumber | 209-702-3 |
| RTECSNumber | XS7965400 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only