missing translation for 'onlineSavingsMsg'
Learn More

3-Bromotoluene, 98%

CAS: 591-17-3 | C7H7Br | 171.04 g/mol

Supplier:  Thermo Scientific Chemicals A1399022

 View more versions of this product

Catalog No. AAA1399022


Only null left
Explore available promotions

Description

Description

3-Bromotoluene was used in the precise determination of bromine isotope ratio in organic compounds by MC-ICPMS ( Multicollector-Inductively Coupled Plasma Mass Spectrometer). It undergoes palladium-catalyzed cyanation reaction in the presence of K4[Fe(CN)6] as the cyanide surrogate. It also undergoes palladium-catalyzed reaction with alkynyltriarylborates to yield trisubstituted alkenylboranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromotoluene was used in the precise determination of bromine isotope ratio in organic compounds by MC-ICPMS ( Multicollector-Inductively Coupled Plasma Mass Spectrometer). It undergoes palladium-catalyzed cyanation reaction in the presence of K4[Fe(CN)6] as the cyanide surrogate. It also undergoes palladium-catalyzed reaction with alkynyltriarylborates to yield trisubstituted alkenylboranes.

Solubility
Not miscible in water. Soluble in alcohols, ether and benzene.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

3-Bromotoluene
-40°C
183°C to 184°C
C7H7Br
MFCD00000085
1903633
3-bromotoluene, m-bromotoluene, 3-methylbromobenzene, benzene, 1-bromo-3-methyl, m-tolyl bromide, toluene, m-bromo, 5-bromotoluene, 3-bromo-1-methylbenzene, m-methylbromobenzene, 3-methyl-1-bromobenzene
WJIFKOVZNJTSGO-UHFFFAOYSA-N
1-bromo-3-methylbenzene
11560
98%
591-17-3
1.41
75°C (167°F)
1.552
100 g
14,1439
Not miscible in water. Soluble in alcohols,ether and benzene.
CC1=CC=CC(Br)=C1
171.04
171.04

Chemical Identifiers

591-17-3
171.04
WJIFKOVZNJTSGO-UHFFFAOYSA-N
11560
CC1=CC=CC(Br)=C1
C7H7Br
MFCD00000085
3-bromotoluene, m-bromotoluene, 3-methylbromobenzene, benzene, 1-bromo-3-methyl, m-tolyl bromide, toluene, m-bromo, 5-bromotoluene, 3-bromo-1-methylbenzene, m-methylbromobenzene, 3-methyl-1-bromobenzene
1-bromo-3-methylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302-H315-H319-H335

DOTInformation : Hazard Class: 3; Packaging Group: III

EINECSNumber : 209-702-3

RTECSNumber : XS7965400

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only