missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromostyrene (stabilized with TBC) 97.0+%, TCI America™
$80.16 - $212.51
Chemical Identifiers
| CAS | 2039-86-3 |
|---|---|
| Molecular Formula | C8H7Br |
| Molecular Weight (g/mol) | 183.048 |
| MDL Number | MFCD00000088 |
| InChI Key | KQJQPCJDKBKSLV-UHFFFAOYSA-N |
| Synonym | 3-bromostyrene, 1-bromo-3-vinylbenzene, m-bromostyrene, benzene, 1-bromo-3-ethenyl, 1-bromo-3-vinyl-benzene, unii-6xe3sf241y, 3-bromo-styrene, pubchem23859, acmc-1cmdc, 1-bromo-3-vinylbenzene # |
| PubChem CID | 74870 |
| IUPAC Name | 1-bromo-3-ethenylbenzene |
| SMILES | C=CC1=CC(=CC=C1)Br |
Chemical Identifiers
| 2039-86-3 | |
| 183.048 | |
| KQJQPCJDKBKSLV-UHFFFAOYSA-N | |
| 74870 | |
| C=CC1=CC(=CC=C1)Br |
| C8H7Br | |
| MFCD00000088 | |
| 3-bromostyrene, 1-bromo-3-vinylbenzene, m-bromostyrene, benzene, 1-bromo-3-ethenyl, 1-bromo-3-vinyl-benzene, unii-6xe3sf241y, 3-bromo-styrene, pubchem23859, acmc-1cmdc, 1-bromo-3-vinylbenzene # | |
| 1-bromo-3-ethenylbenzene |
Specifications
| 2039-86-3 | |
| 94°C | |
| MFCD00000088 | |
| 3-bromostyrene, 1-bromo-3-vinylbenzene, m-bromostyrene, benzene, 1-bromo-3-ethenyl, 1-bromo-3-vinyl-benzene, unii-6xe3sf241y, 3-bromo-styrene, pubchem23859, acmc-1cmdc, 1-bromo-3-vinylbenzene # | |
| C=CC1=CC(=CC=C1)Br | |
| 183.048 | |
| 183.05 | |
| Liquid |
| Yellow | |
| C8H7Br | |
| 1 g | |
| KQJQPCJDKBKSLV-UHFFFAOYSA-N | |
| 1-bromo-3-ethenylbenzene | |
| 74870 | |
| ≥97.0% (GC) | |
| 3-Bromostyrene (stabilized with TBC) |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator