missing translation for 'onlineSavingsMsg'
Learn More

3-Bromostyrene, 97%, stabilized

Catalog No. AC306480010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC306480010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC306480010 Supplier Thermo Scientific Chemicals Supplier No. 306480010

CAS: 2039-86-3 | C8H7Br | 183.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2039-86-3
Molecular Formula C8H7Br
Molecular Weight (g/mol) 183.05
InChI Key KQJQPCJDKBKSLV-UHFFFAOYSA-N
Synonym 3-bromostyrene, 1-bromo-3-vinylbenzene, m-bromostyrene, benzene, 1-bromo-3-ethenyl, 1-bromo-3-vinyl-benzene, unii-6xe3sf241y, 3-bromo-styrene, pubchem23859, acmc-1cmdc, 1-bromo-3-vinylbenzene #
PubChem CID 74870
IUPAC Name 1-bromo-3-ethenylbenzene
SMILES C=CC1=CC(=CC=C1)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4060g/mL
Boiling Point 74.0°C to 75.0°C (3.0 mmHg)
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHC6H4Br
Refractive Index 1.5890 to 1.5910
Quantity 1 g
Specific Gravity 1.406
Stabilizer 0.05% 3, 5-di-tert.-butylcatechol
Formula Weight 183.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Bromostyrene, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 218-025-2

RUO – Research Use Only