missing translation for 'onlineSavingsMsg'
Learn More

3-Bromostyrene, 97%, stab. with 0.1% 4-tert-butylcatechol

CAS: 2039-86-3 | C8H7Br | 183.048 g/mol

$82.08 - $1082.14

Chemical Identifiers

CAS 2039-86-3
Molecular Formula C8H7Br
Molecular Weight (g/mol) 183.048
MDL Number MFCD00000088
InChI Key KQJQPCJDKBKSLV-UHFFFAOYSA-N
Synonym 3-bromostyrene, 1-bromo-3-vinylbenzene, m-bromostyrene, benzene, 1-bromo-3-ethenyl, 1-bromo-3-vinyl-benzene, unii-6xe3sf241y, 3-bromo-styrene, pubchem23859, acmc-1cmdc, 1-bromo-3-vinylbenzene #
PubChem CID 74870
IUPAC Name 1-bromo-3-ethenylbenzene
SMILES C=CC1=CC(=CC=C1)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2449703
View Documents
Thermo Scientific Chemicals
B2449703
1 g
Each for $82.08
Only null left
 
AAB2449706
View Documents
Thermo Scientific Chemicals
B2449706
5 g
Each for $282.45
Only null left
 
AAB2449714
View Documents
Thermo Scientific Chemicals
B2449714
25 g
Each for $1,082.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2039-86-3
183.048
KQJQPCJDKBKSLV-UHFFFAOYSA-N
74870
C=CC1=CC(=CC=C1)Br
C8H7Br
MFCD00000088
3-bromostyrene, 1-bromo-3-vinylbenzene, m-bromostyrene, benzene, 1-bromo-3-ethenyl, 1-bromo-3-vinyl-benzene, unii-6xe3sf241y, 3-bromo-styrene, pubchem23859, acmc-1cmdc, 1-bromo-3-vinylbenzene #
1-bromo-3-ethenylbenzene

Specifications

2039-86-3
73°C to 74°C (3 mmHg)
C8H7Br
MFCD00000088
2038490
KQJQPCJDKBKSLV-UHFFFAOYSA-N
1-bromo-3-ethenylbenzene
74870
97%
1.404
67°C (152°F)
1.591
1 g
3-bromostyrene, 1-bromo-3-vinylbenzene, m-bromostyrene, benzene, 1-bromo-3-ethenyl, 1-bromo-3-vinyl-benzene, unii-6xe3sf241y, 3-bromo-styrene, pubchem23859, acmc-1cmdc, 1-bromo-3-vinylbenzene #
C=CC1=CC(=CC=C1)Br
183.048
183.05
3-Bromostyrene, stabilized with 0.1% 4-tert-butylcatechol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 218-025-2

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only