missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropionitrile, 97%

Catalog No. AAB2399814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2399814 25 g
AAB2399822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2399814 Supplier Thermo Scientific Chemicals Supplier No. B2399814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2417-90-5 | C3H4BrN | 133.976 g/mol

3-Bromopropionitrile was used as the initiator in the atom transfer radical polymerization of acrylonitrile. It was used in coupling reactions of a number of aromatic and heteroaromatic phenols with alkyl, acyl or benzoyl halides in acetonitrile with cesium fluoride-Celite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromopropionitrile was used as the initiator in the atom transfer radical polymerization of acrylonitrile. It was used in coupling reactions of a number of aromatic and heteroaromatic phenols with alkyl, acyl or benzoyl halides in acetonitrile with cesium fluoride-Celite.

Solubility
Soluble in ethanol, ether.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2417-90-5
Molecular Formula C3H4BrN
Molecular Weight (g/mol) 133.976
MDL Number MFCD00001951
InChI Key CQZIEDXCLQOOEH-UHFFFAOYSA-N
Synonym 3-bromopropionitrile, propanenitrile, 3-bromo, 2-bromoethyl cyanide, 3-bromopropiononitrile, 1-bromo-2-cyanoethane, beta-bromopropionitrile, 2-cyanoethyl bromide, propionitrile, 3-bromo, usaf do-51, beta-bromopriopionitrile
PubChem CID 17020
IUPAC Name 3-bromopropanenitrile
SMILES C(CBr)C#N

Specifications

Density 1.615
Boiling Point 200°C
Flash Point 97°C (206°F)
Assay Percent Range 0.98
Refractive Index 1.481
Quantity 25 g
UN Number UN3276
Beilstein 1738565
Solubility Information Soluble in ethanol,ether.
Formula Weight 133.98
Percent Purity 98%
Chemical Name or Material 3-Bromopropionitrile
Show More Show Less
Hazard Category H301-H315-H319
Hazard Statement GHS H Statement
H300-H310-H330-H315-H319-H335
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 219-332-4
RTECSNumber UG1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only