accessibility menu, dialog, popup

UserName

BrN

CAS RN 2417-90-5

IUPAC Name: 3-bromopropanenitrile
Synonyms: propanenitrile, 3-bromo-
Molecular Weight (g/mol): 133.98
Molecular Formula: C3H4BrN
InChi Key: CQZIEDXCLQOOEH-UHFFFAOYSA-N
SMILES: BrCCC#N
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)