missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropionic Acid 98.0+%, TCI America™

Supplier:  TCI America B0645500G

 View more versions of this product

Catalog No. B0645500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Bromopropionic Acid
63°C
142°C
MFCD00002763
3261
DHXNZYCXMFBMHE-UHFFFAOYSA-N
3-bromopropanoic acid
11553
≥98.0% (GC,T)
590-92-1
Red-White
C3H5BrO2
500 g
3-bromopropionic acid, propanoic acid, 3-bromo, beta-bromopropionic acid, propionic acid, 3-bromo, 2-carboxyethyl bromide, beta-bromopropanoic acid, unii-wfz7csr69r, beta-propionic acid, 3-bromo-propionic acid, wfz7csr69r
OC(=O)CCBr
152.98
152.98
Crystalline Powder

Chemical Identifiers

590-92-1
152.98
DHXNZYCXMFBMHE-UHFFFAOYSA-N
11553
OC(=O)CCBr
C3H5BrO2
MFCD00002763
3-bromopropionic acid, propanoic acid, 3-bromo, beta-bromopropionic acid, propionic acid, 3-bromo, 2-carboxyethyl bromide, beta-bromopropanoic acid, unii-wfz7csr69r, beta-propionic acid, 3-bromo-propionic acid, wfz7csr69r
3-bromopropanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3937

RTECSNumber : UE7875000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions