accessibility menu, dialog, popup

UserName

OHOBr

CAS RN 590-92-1

IUPAC Name: 3-bromopropanoic acid
Synonyms: propanoic acid, 3-bromo-
Molecular Weight (g/mol): 152.98
Molecular Formula: C3H5BrO2
InChi Key: DHXNZYCXMFBMHE-UHFFFAOYSA-N
SMILES: OC(=O)CCBr
Boiling Point 
  • (3)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)