missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropionic Acid 98.0+%, TCI America™

Catalog No. B064525G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B064525G 25 g
B0645100G 100 g
B0645500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B064525G Supplier TCI America Supplier No. B064525G

Chemical Identifiers

CAS 590-92-1
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.98
MDL Number MFCD00002763
InChI Key DHXNZYCXMFBMHE-UHFFFAOYSA-N
Synonym 3-bromopropionic acid, propanoic acid, 3-bromo, beta-bromopropionic acid, propionic acid, 3-bromo, 2-carboxyethyl bromide, beta-bromopropanoic acid, unii-wfz7csr69r, beta-propionic acid, 3-bromo-propionic acid, wfz7csr69r
PubChem CID 11553
IUPAC Name 3-bromopropanoic acid
SMILES OC(=O)CCBr

Specifications

Melting Point 63°C
Color Red-White
Boiling Point 142°C
Quantity 25 g
UN Number 3261
Formula Weight 152.98
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromopropionic Acid
EINECSNumber (2)-3937
RTECSNumber UE7875000
TSCA Yes