missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropionic acid, 97%

Catalog No. AAA1157314
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1157314 25 g
AAA1157322 100 g
AAA1157336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1157314 Supplier Thermo Scientific Chemicals Supplier No. A1157314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 590-92-1 | C3H5BrO2 | 152.98 g/mol

3-Bromopropionic acid used as quaternization agent in amperometric biosensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromopropionic acid used as quaternization agent in amperometric biosensors.

Solubility
Soluble in water, alcohol, ether, chloroform, benzene

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 590-92-1
Molecular Formula C3H5BrO2
Molecular Weight (g/mol) 152.98
MDL Number MFCD00002763
InChI Key DHXNZYCXMFBMHE-UHFFFAOYSA-N
Synonym 3-bromopropionic acid, propanoic acid, 3-bromo, beta-bromopropionic acid, propionic acid, 3-bromo, 2-carboxyethyl bromide, beta-bromopropanoic acid, unii-wfz7csr69r, beta-propionic acid, 3-bromo-propionic acid, wfz7csr69r
PubChem CID 11553
IUPAC Name 3-bromopropanoic acid
SMILES OC(=O)CCBr

Specifications

Melting Point 57°C to 63°C
Density 1.48
Boiling Point 140°C to 142°C (45 mmHg)
Flash Point 65°C (149°F)
Quantity 25 g
UN Number UN3261
Beilstein 1071333
Merck Index 14,1433
Solubility Information Soluble in water, alcohol, ether, chloroform, benzene
Formula Weight 152.98
Percent Purity 97%
Chemical Name or Material 3-Bromopropionic acid
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 209-694-1
RTECSNumber UE7875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only