missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Bromophenyl)thiourea 98.0+%, TCI America™
$106.25 - $354.17
Chemical Identifiers
| CAS | 21327-14-0 |
|---|---|
| Molecular Formula | C7H7BrN2S |
| Molecular Weight (g/mol) | 231.111 |
| MDL Number | MFCD00060440 |
| InChI Key | XBRVSIPVHYWULW-UHFFFAOYSA-N |
| PubChem CID | 2735619 |
| IUPAC Name | (3-bromophenyl)thiourea |
| SMILES | C1=CC(=CC(=C1)Br)NC(=S)N |
Chemical Identifiers
| 21327-14-0 | |
| 231.111 | |
| XBRVSIPVHYWULW-UHFFFAOYSA-N | |
| (3-bromophenyl)thiourea |
| C7H7BrN2S | |
| MFCD00060440 | |
| 2735619 | |
| C1=CC(=CC(=C1)Br)NC(=S)N |
Specifications
| 21327-14-0 | |
| White-Yellow | |
| MFCD00060440 | |
| 2811 | |
| C1=CC(=CC(=C1)Br)NC(=S)N | |
| 231.111 | |
| 231.11 | |
| Crystalline Powder |
| 147°C | |
| C7H7BrN2S | |
| 5 g | |
| XBRVSIPVHYWULW-UHFFFAOYSA-N | |
| (3-bromophenyl)thiourea | |
| 2735619 | |
| ≥98.0% (HPLC,N) | |
| (3-Bromophenyl)thiourea |
Safety and Handling
TSCA : No