accessibility menu, dialog, popup

UserName

CAS RN 21327-14-0

IUPAC Name: (3-bromophenyl)thiourea
Synonyms: 3-bromophenylthiourea
Molecular Weight (g/mol): 231.11
Molecular Formula: C7H7BrN2S
InChi Key: XBRVSIPVHYWULW-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=CC(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results