missing translation for 'onlineSavingsMsg'
Learn More

(3-Bromophenyl)thiourea 98.0+%, TCI America™

Catalog No. B49985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49985G 5 g
B499825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49985G Supplier TCI America Supplier No. B49985G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21327-14-0
Molecular Formula C7H7BrN2S
Molecular Weight (g/mol) 231.111
MDL Number MFCD00060440
InChI Key XBRVSIPVHYWULW-UHFFFAOYSA-N
PubChem CID 2735619
IUPAC Name (3-bromophenyl)thiourea
SMILES C1=CC(=CC(=C1)Br)NC(=S)N

Specifications

Melting Point 147°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 231.11
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (3-Bromophenyl)thiourea