missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenyl isothiocyanate, 97%

Catalog No. p-7045224
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0987106 5 g
AAL0987114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0987106 Supplier Thermo Scientific Chemicals Supplier No. L0987106

CAS: 2131-59-1 | C7H4BrNS | 214.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2131-59-1
Molecular Formula C7H4BrNS
Molecular Weight (g/mol) 214.08
MDL Number MFCD00004803
InChI Key ZMGMGHNOACSMQN-UHFFFAOYSA-N
Synonym 3-bromophenyl isothiocyanate, m-bromophenyl isothiocyanate, benzene, 1-bromo-3-isothiocyanato, isothiocyanic acid, m-bromophenyl ester, 3-bromophenylisothiocyanate, 1-bromo-3-isothiocyanato-benzene, acmc-1chhg, m-bromphenylisothiocyanat, zmgmghnoacsmqn-uhfffaoysa, #
PubChem CID 16481
IUPAC Name 1-bromo-3-isothiocyanatobenzene
SMILES C1=CC(=CC(=C1)Br)N=C=S

Specifications

Boiling Point 255°C to 256°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.685
Quantity 5 g
UN Number UN2922
Beilstein 2081173
Sensitivity Moisture sensitive
Formula Weight 214.09
Percent Purity 97%
Chemical Name or Material 3-Bromophenyl isothiocyanate
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
RTECSNumber NX8350000
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only