accessibility menu, dialog, popup

UserName

BrNCS

CAS RN 2131-59-1

IUPAC Name: 1-bromo-3-isothiocyanatobenzene
Synonyms: 1-bromo-3-isothiocyanatobenzene
Molecular Weight (g/mol): 214.08
Molecular Formula: C7H4BrNS
InChi Key: ZMGMGHNOACSMQN-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)N=C=S
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)