missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenyl Isocyanate 98.0+%, TCI America™

Catalog No. p-7131480
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05535G 5 g
I055325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05535G Supplier TCI America Supplier No. I05535G

Chemical Identifiers

CAS 23138-55-8
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.02
MDL Number MFCD00002014
InChI Key VQVBCZQTXSHJGF-UHFFFAOYSA-N
Synonym 3-bromophenyl isocyanate, 3-bromophenylisocyanate, benzene, 1-bromo-3-isocyanato, isocyanic acid 3-bromophenyl ester, 3-bromobenzenisocyanate, m-bromophenylisocyanate, 3-bromophenyliso-cyanate, 3-bromo phenyl isocyanate, 3-bromo-phenyl isocyanate, acmc-209g1f
PubChem CID 141001
IUPAC Name 1-bromo-3-isocyanatobenzene
SMILES BrC1=CC=CC(=C1)N=C=O

Specifications

Color Yellow
Boiling Point 220°C
Quantity 5 g
UN Number 2206
Formula Weight 198.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromophenyl Isocyanate
TSCA No
Recommended Storage Refrigerator