missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenol 95.0+%, TCI America™

Catalog No. B062925G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B062925G 25 g
B0629100G 100 g
B0629500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B062925G Supplier TCI America Supplier No. B062925G

Chemical Identifiers

CAS 591-20-8
Molecular Formula C6H5BrO
Molecular Weight (g/mol) 173.01
MDL Number MFCD00002253
InChI Key MNOJRWOWILAHAV-UHFFFAOYSA-N
Synonym m-bromophenol, phenol, 3-bromo, 3-bromo phenol, 3-bromo-phenol, phenol, m-bromo, unii-vmu0x6956y, meta bromophenol, meta-bromophenol, 3 bromophenol, 3-bromanylphenol
PubChem CID 11563
IUPAC Name 3-bromophenol
SMILES OC1=CC=CC(Br)=C1

Specifications

Color Yellow
Boiling Point 237°C
Quantity 25 g
Formula Weight 173.01
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Bromophenol
EINECSNumber (3)-0896
TSCA Yes